accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 64920-29-2

IUPAC Name: ethyl 2-oxo-4-phenylbutanoate
Synonyms: ethyl 2-oxo-4-phenylbutanoate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: STPXIOGYOLJXMZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results