CAS RN 6485-97-8
IUPAC Name:
2-phenyl-9,10-dihydroanthracene-9,10-dione
Synonyms:
2-phenylanthracene-9,10-dione
Molecular Weight (g/mol):
284.31
Molecular Formula:
C20H12O2
InChi Key:
NTZCFGZBDDCNHI-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2C(=O)C2=CC(=CC=C12)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Phenylanthraquinone 98.0+%, TCI America™
CAS: 6485-97-8 Molecular Formula: C20H12O2 Molecular Weight (g/mol): 284.314 MDL Number: MFCD00559262 InChI Key: NTZCFGZBDDCNHI-UHFFFAOYSA-N Synonym: 2-phenylanthraquinone,2-phenylanthra-9,10-quinone,2-phenyl-9,10-anthracenedione,2-phenylanthrachinon,acmc-20annp,9, 2-phenyl,2-phenyl-9,10-anthraquinone,9,10-anthracenedione,2-phenyl PubChem CID: 313995 IUPAC Name: 2-phenylanthracene-9,10-dione SMILES: C1=CC=C(C=C1)C2=CC3=C(C=C2)C(=O)C4=CC=CC=C4C3=O