missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylanthraquinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 162°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 284.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Phenylanthraquinone |
Safety and Handling
| TSCA | No |