accessibility menu, dialog, popup

UserName

H3CNHNClCl

CAS RN 6478-79-1

IUPAC Name: 5,6-dichloro-2-methyl-1H-1,3-benzodiazole
Synonyms: 5,6-dichloro-2-methyl-1H-1,3-benzodiazole
Molecular Weight (g/mol): 201.05
Molecular Formula: C8H6Cl2N2
InChi Key: WMOBNOCVMZFPEN-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(Cl)=C(Cl)C=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results