accessibility menu, dialog, popup

UserName

CAS RN 64485-88-7

IUPAC Name: ethyl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Synonyms: ethyl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Molecular Weight (g/mol): 229.25
Molecular Formula: C8H11N3O3S
InChi Key: POBMBNPEUPDXRS-WDZFZDKYSA-N
SMILES: CCOC(=O)C(=N/OC)\C1=CSC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results