accessibility menu, dialog, popup

UserName

H3COOCH2OO

CAS RN 6439-57-2

IUPAC Name: 3-(butoxycarbonyl)but-3-enoate
Synonyms: 3-(butoxycarbonyl)but-3-enoate
Molecular Weight (g/mol): 185.2
Molecular Formula: C9H13O4
InChi Key: DYYFCJRYZVHEKQ-UHFFFAOYSA-M
SMILES: CCCCOC(=O)C(=C)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)