accessibility menu, dialog, popup

UserName

CAS RN 6437-49-6

IUPAC Name: 2-phenoxypropan-1-amine hydrochloride
Synonyms: 2-phenoxypropan-1-amine hydrochloride
Molecular Weight (g/mol): 187.67
Molecular Formula: C9H14ClNO
InChi Key: CGOBBGUREQJPPH-UHFFFAOYNA-N
SMILES: Cl.CC(CN)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results