accessibility menu, dialog, popup

UserName

CAS RN 64277-87-8

CH3H3CCH3OOH3CCH3H3CCH3

CAS RN 64277-87-8

IUPAC Name: methyl 3,5-di-tert-butylbenzoate
Synonyms: methyl 3,5-di-tert-butylbenzoate
Molecular Weight (g/mol): 248.37
Molecular Formula: C16H24O2
InChi Key: ZEIOQJMJXFVPOG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results