missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Di-tert-butylbenzoate 98.0+%, TCI America™

Catalog No. D26955G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26955G Supplier TCI America Supplier No. D26955G
Only null left

Chemical Identifiers

CAS 64277-87-8
Molecular Formula C16H24O2
Molecular Weight (g/mol) 248.37
MDL Number MFCD03844786
InChI Key ZEIOQJMJXFVPOG-UHFFFAOYSA-N
Synonym 3,5-Di-tert-butylbenzoic Acid Methyl Ester
PubChem CID 13267559
IUPAC Name methyl 3,5-di-tert-butylbenzoate
SMILES COC(=O)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 53°C
Color White
Quantity 5 g
Formula Weight 248.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Di-tert-butylbenzoate