accessibility menu, dialog, popup

UserName

CAS RN 64228-81-5

IUPAC Name: 1-[(3,4-dimethoxyphenyl)methyl]-2-[3-({5-[(3-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium-2-yl}propanoyl)oxy]pentyl}oxy)-3-oxopropyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium dibenzenesulfonate
Synonyms: atracurium besylate atracurii besilas besilate d'atracurium 苯磺阿曲库铵 atracurium besilate besilato de atracurio
Molecular Weight (g/mol): 1243.49
Molecular Formula: C65H82N2O18S2
InChi Key: XXZSQOVSEBAPGS-UHFFFAOYNA-L
SMILES: [O-]S(=O)(=O)C1=CC=CC=C1.[O-]S(=O)(=O)C1=CC=CC=C1.COC1=CC=C(CC2C3=CC(OC)=C(OC)C=C3CC[N+]2(C)CCC(=O)OCCCCCOC(=O)CC[N+]2(C)CCC3=CC(OC)=C(OC)C=C3C2CC2=CC=C(OC)C(OC)=C2)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results