accessibility menu, dialog, popup

UserName

H3C(R)OHONH2CH3

CAS RN 640-68-6

IUPAC Name: (2R)-2-amino-3-methylbutanoic acid
Synonyms: D-valine|D-valin
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: KZSNJWFQEVHDMF-SCSAIBSYSA-N
SMILES: CC(C)[C@@H](N)C(O)=O
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)