accessibility menu, dialog, popup

UserName

NaHO(R)O(R)OHOOOH

CAS RN 6381-77-7

IUPAC Name: sodium (2R)-2-[(2R)-4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl]-2-hydroxyethan-1-olate
Synonyms: sodium erythorbate
Molecular Weight (g/mol): 198.11
Molecular Formula: C6H7NaO6
InChi Key: QNSCGSPRKIPKTN-RKJRWTFHSA-N
SMILES: [Na+].O[C@H](C[O-])[C@H]1OC(O)=C(O)C1=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)