CAS RN 637-50-3
IUPAC Name:
[(1E)-prop-1-en-1-yl]benzene
Synonyms:
trans-1-phenyl-1-propene
Molecular Weight (g/mol):
118.18
Molecular Formula:
C9H10
InChi Key:
QROGIFZRVHSFLM-QHHAFSJGSA-N
SMILES:
C\C=C\C1=CC=CC=C1
1
–
1
of
1
results
1
beta-Methylstyrene (cis- and trans- mixture) (stabilized with TBC) 95.0+%, TCI America™
CAS: 637-50-3 Molecular Formula: C9H10 Molecular Weight (g/mol): 118.18 MDL Number: MFCD00009280 InChI Key: QROGIFZRVHSFLM-QHHAFSJGSA-N Synonym: beta-methylstyrene,trans-beta-methylstyrene,1-phenylpropene,isoallylbenzene,propenylbenzene,trans-1-phenyl-1-propene,1-propenylbenzene,trans-propenylbenzene,1-phenyl-1-propene,benzene, 1-propenyl PubChem CID: 252325 IUPAC Name: [(1E)-prop-1-en-1-yl]benzene SMILES: C\C=C\C1=CC=CC=C1