missing translation for 'onlineSavingsMsg'
Learn More

beta-Methylstyrene (cis- and trans- mixture) (stabilized with TBC) 95.0+%, TCI America™

Catalog No. P049525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P049525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P049525ML Supplier TCI America Supplier No. P049525ML
Only null left

Chemical Identifiers

CAS 637-50-3
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009280
InChI Key QROGIFZRVHSFLM-QHHAFSJGSA-N
Synonym beta-methylstyrene, trans-beta-methylstyrene, 1-phenylpropene, isoallylbenzene, propenylbenzene, trans-1-phenyl-1-propene, 1-propenylbenzene, trans-propenylbenzene, 1-phenyl-1-propene, benzene, 1-propenyl
PubChem CID 252325
IUPAC Name [(1E)-prop-1-en-1-yl]benzene
SMILES C\C=C\C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
UN Number 3295
Formula Weight 118.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Methylstyrene (cis- and trans- mixture) (stabilized with TBC)
EINECSNumber (3)-0008
RTECSNumber DA8400000
TSCA No