accessibility menu, dialog, popup

UserName

H3CCH3CH3OONH(R)OHOO

CAS RN 6368-20-3

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoate
Molecular Weight (g/mol): 204.2
Molecular Formula: C8H14NO5
InChi Key: FHOAKXBXYSJBGX-RXMQYKEDSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CO)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)