accessibility menu, dialog, popup

UserName

CAS RN 63659-19-8

IUPAC Name: hydrogen 1-{4-[2-(cyclopropylmethoxy)ethyl]phenoxy}-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms: hydrogen betaxolol chloride
Molecular Weight (g/mol): 343.89
Molecular Formula: C18H30ClNO3
InChi Key: CHDPSNLJFOQTRK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)NCC(O)COC1=CC=C(CCOCC2CC2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)