missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Betaxolol hydrochloride

Numéro de catalogue. AC462440010
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC462440010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC462440010 Fournisseur Thermo Scientific™ Code fournisseur 462440010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 63659-19-8
Formule moléculaire C18H30ClNO3
Poids moléculaire (g/mol) 343.89
Clé InChI CHDPSNLJFOQTRK-UHFFFAOYNA-N
Nom de l’IUPAC hydrogen 1-{4-[2-(cyclopropylmethoxy)ethyl]phenoxy}-3-[(propan-2-yl)amino]propan-2-ol chloride
SOURIRES [H+].[Cl-].CC(C)NCC(O)COC1=CC=C(CCOCC2CC2)C=C1

Spécifications

Nom ou substance chimique βxolol hydrochloride
Quantité 1 g
Renseignements sur la solubilité Solubility aq. soln.: very soluble aq. soln.. Other solubilities: very soluble in methanol and ethanol, slightly soluble in acetone and acetonitrile, insoluble in ether
Poids de formule 343.89
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 264-384-3

RUO – Research Use Only