accessibility menu, dialog, popup

UserName

OOHOHOOHOOHOHHOHOOHH2O

CAS RN 6363-53-7

IUPAC Name: 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Synonyms: cellobiose hydrate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: HBDJFVFTHLOSDW-UHFFFAOYNA-N
SMILES: O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (5)
  • (3)
  • (3)
  • (1)
Grade 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)