missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose Monohydrate, Fisher BioReagents

Catalog No. BP684500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP684500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP684500 Supplier Fisher BioReagents Supplier No. FLBP684 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maltose is suitable for use in cell culture systems requiring sugar additives.

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Alpha Vector Maltose Monohydrate
Melting Point 130°C
Color Colorless
pH 5.5
Loss on Drying record%
Assay Percent Range ≥97 %
Packaging Poly Bottle
Quantity 500 g
Specific Rotation 128 to 132° (+ or -)
Formula Weight 360.29
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material D-(+)-Maltose Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT