missing translation for 'onlineSavingsMsg'
Learn More

Maltose, Monohydrate, NF, 92%, Spectrum™ Chemical

Catalog No. 18604174
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604174 12 kg Poly Bottle
18604172 500 g Poly Bottle
18604173 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604174 Supplier Spectrum Chemical Mfg Cor Supplier No. M107912KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1079, 6363-53-7, C12H22O11·H2O

Spectrum™ Chemical Maltose, Monohydrate, NF is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Poly Bottle
Quantity 12 kg
Percent Purity 92%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Maltose, Monohydrate