accessibility menu, dialog, popup

UserName

CH3H3CCH2

CAS RN 6362-80-7

IUPAC Name: (4-methyl-4-phenylpent-1-en-2-yl)benzene
Synonyms: (4-methyl-4-phenylpent-1-en-2-yl)benzene
Molecular Weight (g/mol): 236.36
Molecular Formula: C18H20
InChi Key: ZOKCNEIWFQCSCM-UHFFFAOYSA-N
SMILES: CC(C)(CC(=C)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results