accessibility menu, dialog, popup

UserName

CAS RN 6359-98-4

IUPAC Name: disodium 2,5-dichloro-4-{3-methyl-5-oxo-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazol-1-yl}benzene-1-sulfonate
Synonyms: yellow 2G
Molecular Weight (g/mol): 551.28
Molecular Formula: C16H10Cl2N4Na2O7S2
InChi Key: FTZLWXQKVFFWLY-UHFFFAOYNA-L
SMILES: [Na+].[Na+].CC1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC(Cl)=C(C=C1Cl)S([O-])(=O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results