missing translation for 'onlineSavingsMsg'
Learn More

Yellow 2G, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014998
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014998 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014998 Supplier Merck Emd Millipore Supplier No. 0029825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 6359-98-4
Linear Formula C16H10 Cl2N4 Na2O7S2
MDL Number MFCD00053367
Synonym Acid Yellow 17
Type Analytical Standard
Quantity 25 mg
Formula Weight 551.29
Percent Purity ≥98% (HPLC)
Grade Analytical Standard