missing translation for 'onlineSavingsMsg'
Learn More

Xylene Fast Yellow 2G, TCI America™

Catalog No. X002525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X002525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X002525G Supplier TCI America Supplier No. X002525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6359-98-4
Molecular Formula C16H12Cl2N4Na2O7S2
Molecular Weight (g/mol) 553.293
MDL Number MFCD00053367
InChI Key WXBJLALITVDPLG-UHFFFAOYSA-N
Synonym c.i.acid yellow 17, yellow 2g, analytical standard, acid yellow 17, dye content 60 %
PubChem CID 87601076
IUPAC Name 2,5-dichloro-4-[3-methyl-5-oxo-4-[(4-sulfophenyl)diazenyl]-4H-pyrazol-1-yl]benzenesulfonic acid;sodium
SMILES CC1=NN(C(=O)C1N=NC2=CC=C(C=C2)S(=O)(=O)O)C3=CC(=C(C=C3Cl)S(=O)(=O)O)Cl.[Na].[Na]

Specifications

Quantity 25 g
Formula Weight 551.28
Physical Form Crystalline Powder
Chemical Name or Material Xylene Fast Yellow 2G
EINECSNumber (5)-1399
TSCA Yes