accessibility menu, dialog, popup

UserName

CAS RN 6336-51-2

IUPAC Name: 3-phenyl-1,3,4-thiadiazolidine-2,5-dithione potassium
Synonyms: 3-phenyl-1,3,4-thiadiazolidine-2,5-dithione potassium
Molecular Weight (g/mol): 265.43
Molecular Formula: C8H6KN2S3
InChi Key: UNERKJMVFVZPDY-UHFFFAOYSA-N
SMILES: [K].S=C1NN(C(=S)S1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)