accessibility menu, dialog, popup

UserName

CAS RN 632-15-5

IUPAC Name: 3,4-dimethylthiophene
Synonyms: 3,4-dimethylthiophene
Molecular Weight (g/mol): 112.19
Molecular Formula: C6H8S
InChi Key: GPSFYJDZKSRMKZ-UHFFFAOYSA-N
SMILES: CC1=CSC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results