accessibility menu, dialog, popup

UserName

CAS RN 6318-56-5

IUPAC Name: 5-aminobenzene-1,3-diol hydrochloride
Synonyms: 5-aminobenzene-1,3-diol hydrochloride
Molecular Weight (g/mol): 161.59
Molecular Formula: C6H8ClNO2
InChi Key: VNZZCDQPCQIUGG-UHFFFAOYSA-N
SMILES: Cl.NC1=CC(O)=CC(O)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)