missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoveratrole, 98%

Catalog No. AC104720250
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104720250 25 g Glass bottle
AC104720100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104720250 Supplier Thermo Scientific Chemicals Supplier No. 104720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6315-89-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008394
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
Synonym 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
PubChem CID 22770
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=C(C=C(C=C1)N)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 88°C
Color Brown
Boiling Point 174°C to 176°C (22.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H3(OCH3)2
Quantity 25 g
Beilstein 13, 780
Solubility Information Solubility in water: insoluble
Formula Weight 153.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoveratrole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 228-647-6
RTECSNumber BX4550000
TSCA TSCA

RUO – Research Use Only