missing translation for 'onlineSavingsMsg'
Learn More

Benedicts Reagent, For the Qualitative Determination of Sugars, MilliporeSigma™ Supelco™

Catalog No. 111019535
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019535 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019535 Supplier Merck Emd Millipore Supplier No. 11945250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benedict′s reagents comprise of copper sulphate, sodium carbonate which functions as a mild alkali, and sodium citrate. It is quite stable. On boiling this reagent with glucose, it gives a white precipitate of cuprous thiocyanate. It is primarily used for the estimation of glucose in urine.

Reagent used for the detection of reducing sugars and other reducing substances; detection limit: 0.01% glucose in water

Specifications

CAS 63126-89-6
MDL Number MFCD00146068
Quantity 250 mL