accessibility menu, dialog, popup

UserName

CAS RN 629-82-3

H3COCH3

CAS RN 629-82-3

IUPAC Name: 1-(octyloxy)octane
Synonyms: antar dicaprylyl ether
Molecular Weight (g/mol): 242.45
Molecular Formula: C16H34O
InChi Key: NKJOXAZJBOMXID-UHFFFAOYSA-N
SMILES: CCCCCCCCOCCCCCCCC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results