accessibility menu, dialog, popup

UserName

CAS RN 629-45-8

IUPAC Name: 1-(butyldisulfanyl)butane
Synonyms: dibutyl disulfide
Molecular Weight (g/mol): 178.35
Molecular Formula: C8H18S2
InChi Key: CUDSBWGCGSUXDB-UHFFFAOYSA-N
SMILES: CCCCSSCCCC
Boiling Point 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)