accessibility menu, dialog, popup

UserName

H3COOCH3MgBr

CAS RN 62890-98-6

IUPAC Name: bromo(2,5-dimethoxyphenyl)magnesium
Synonyms: bromo(2,5-dimethoxyphenyl)magnesium
Molecular Weight (g/mol): 241.37
Molecular Formula: C8H9BrMgO2
InChi Key: GRSCPLVZIMBKKY-UHFFFAOYSA-M
SMILES: COC1=CC=C(OC)C([Mg]Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results