accessibility menu, dialog, popup

UserName

CAS RN 62625-22-3

IUPAC Name: sodium (3Z)-3-{[(E)-{[2-(2-carboxyphenyl)hydrazin-1-yl](phenyl)methylidene}amino]imino}-4-oxocyclohexa-1,5-diene-1-sulfonate
Synonyms: sodium (3Z)-3-{[(E)-{[2-(2-carboxyphenyl)hydrazin-1-yl](phenyl)methylidene}amino]imino}-4-oxocyclohexa-1,5-diene-1-sulfonate
Molecular Weight (g/mol): 462.41
Molecular Formula: C20H15N4NaO6S
InChi Key: JCJLYLAKMAERBD-JJECXDOKSA-M
SMILES: [Na+].OC(=O)C1=CC=CC=C1NN\C(=N\N=C1\C=C(C=CC1=O)S([O-])(=O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)