CAS RN 626-27-7
CAS RN 626-27-7
IUPAC Name:
heptanoyl heptanoate
Synonyms:
heptanoic acid, anhydride
Molecular Weight (g/mol):
242.36
Molecular Formula:
C14H26O3
InChi Key:
DAPZDAPTZFJZTO-UHFFFAOYSA-N
SMILES:
CCCCCCC(=O)OC(=O)CCCCCC
1
–
1
of
1
results
1
Heptanoic Anhydride 97.0+%, TCI America™
CAS: 626-27-7 Molecular Formula: C14H26O3 Molecular Weight (g/mol): 242.359 MDL Number: MFCD00009536 InChI Key: DAPZDAPTZFJZTO-UHFFFAOYSA-N Synonym: heptanoic anhydride,enanthic anhydride,heptanoic acid, anhydride,heptanoyl anhydride,n-heptanoic acid anhydride,n-heptanoic anhydride,heptanoic acid, 1,1'-anhydride,oenanthic anhydride,bisenanthic anhydride,enanthic acid anhydride PubChem CID: 69376 IUPAC Name: heptanoyl heptanoate SMILES: CCCCCCC(=O)OC(=O)CCCCCC