accessibility menu, dialog, popup

UserName

NH2I

CAS RN 626-01-7

IUPAC Name: 3-iodoaniline
Synonyms: benzenamine, 3-iodo-
Molecular Weight (g/mol): 219.03
Molecular Formula: C6H6IN
InChi Key: FFCSRWGYGMRBGD-UHFFFAOYSA-N
SMILES: NC1=CC=CC(I)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)