missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoaniline, 98%

Catalog No. AAL0115918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0115918 50 g
AAL0115909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0115918 Supplier Thermo Scientific Chemicals Supplier No. L0115918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-01-7 | C6H6IN | 219.025 g/mol

3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.

Chemical Identifiers

CAS 626-01-7
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007781
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
PubChem CID 12271
IUPAC Name 3-iodoaniline
SMILES C1=CC(=CC(=C1)I)N

Specifications

Melting Point 21°C to 24°C
Density 1.821
Boiling Point 145°C to 146°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.684
Quantity 50 g
Beilstein 636058
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 219.03
Percent Purity 98%
Chemical Name or Material 3-Iodoaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-922-7
RTECSNumber BY3750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only