accessibility menu, dialog, popup

UserName

CAS RN 622405-78-1

H3COOOOOONOO

CAS RN 622405-78-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2,5,8,11-tetraoxatetradecan-14-oate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2,5,8,11-tetraoxatetradecan-14-oate
Molecular Weight (g/mol): 333.34
Molecular Formula: C14H23NO8
InChi Key: MAYFFZZPEREGBQ-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results