accessibility menu, dialog, popup

UserName

NH3(R)OOCl

CAS RN 6212-33-5

IUPAC Name: (2R)-2-azaniumyl-2-(4-chlorophenyl)acetate
Synonyms: (2R)-2-ammonio-2-(4-chlorophenyl)acetate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: QGJGBYXRJVIYGA-SSDOTTSWSA-N
SMILES: [NH3+][C@@H](C([O-])=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)