accessibility menu, dialog, popup

UserName

CAS RN 6211-24-1

IUPAC Name: barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
Synonyms: barium(2+) bis(4-(phenylamino)benzenesulfonate)
Molecular Weight (g/mol): 633.88
Molecular Formula: C24H20BaN2O6S2
InChi Key: IVCNVXFNTKXMCA-UHFFFAOYSA-L
SMILES: [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (5)
  • (5)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (4)
  • (2)
Grade 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (5)
  • (5)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)