accessibility menu, dialog, popup

UserName

CAS RN 621-64-7

IUPAC Name: nitrosodipropylamine
Synonyms: nitrosodipropylamine
Molecular Weight (g/mol): 130.19
Molecular Formula: C6H14N2O
InChi Key: YLKFDHTUAUWZPQ-UHFFFAOYSA-N
SMILES: CCCN(CCC)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results