missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodi-n-propylamine, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Ampule |
| CAS | 621-64-7 |
| Physical Form | Neat |
| MDL Number | MFCD00013891 |
| Type | Food and Cosmetic Component Standard |
| Quantity | 100 mg |
| Grade | Analytical Standard |