accessibility menu, dialog, popup

UserName

H3COOSOOH3C

CAS RN 62020-09-1

IUPAC Name: methyl 2-methanesulfonylacetate
Synonyms: methyl 2-methanesulfonylacetate
Molecular Weight (g/mol): 152.16
Molecular Formula: C4H8O4S
InChi Key: RQKDASVRZONLNQ-UHFFFAOYSA-N
SMILES: COC(=O)CS(C)(=O)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)