accessibility menu, dialog, popup

UserName

CAS RN 62-51-1

ClH3CNH3CH3CCH3OCH3O

CAS RN 62-51-1

IUPAC Name: [2-(acetyloxy)propyl]trimethylazanium chloride
Synonyms: methacholine chloride provocholine 氯醋甲胆碱
Molecular Weight (g/mol): 195.69
Molecular Formula: C8H18ClNO2
InChi Key: JHPHVAVFUYTVCL-UHFFFAOYNA-M
SMILES: [Cl-].CC(C[N+](C)(C)C)OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results