accessibility menu, dialog, popup

UserName

CAS RN 61948-85-4

IUPAC Name: 2-amino-3,4,5-trimethoxybenzoic acid
Synonyms: 2-amino-3,4,5-trimethoxybenzoic acid
Molecular Weight (g/mol): 227.22
Molecular Formula: C10H13NO5
InChi Key: JSHSRQCOCMIIPA-UHFFFAOYSA-N
SMILES: COC1=CC(C(O)=O)=C(N)C(OC)=C1OC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)