accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 6192-44-5

IUPAC Name: 2-phenoxyethyl acetate
Synonyms: ethanol, 2-phenoxy-, acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: WHFKYDMBUMLWDA-UHFFFAOYSA-N
SMILES: CC(=O)OCCOC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results