missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl Acetate 99.0+%, TCI America™

Catalog No. A0935500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0935500G 500 g
A093525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0935500G Supplier TCI America Supplier No. A0935500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6192-44-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00059336
InChI Key WHFKYDMBUMLWDA-UHFFFAOYSA-N
Synonym Acetic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acetate
PubChem CID 22568
IUPAC Name 2-phenoxyethyl acetate
SMILES CC(=O)OCCOC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 231°C
Quantity 500 g
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethyl Acetate
EINECSNumber (3)-0564
RTECSNumber KM0525000
TSCA No