accessibility menu, dialog, popup

UserName

H2NOCl

CAS RN 619-56-7

IUPAC Name: 4-chlorobenzamide
Synonyms: 4-chlorobenzamide
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: BLNVISNJTIRAHF-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)