accessibility menu, dialog, popup

UserName

H3COHHNOCH3OOH3C

CAS RN 616867-28-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2,3-dimethylbutanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: AWEZXIRZNQCCNN-OAQYLSRUSA-N
SMILES: CC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results