accessibility menu, dialog, popup

UserName

CAS RN 61439-59-6

IUPAC Name: 2-[4-(benzyloxy)phenyl]ethan-1-ol
Synonyms: 2-[4-(benzyloxy)phenyl]ethanol
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: JCUJAHLWCDISCC-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results