accessibility menu, dialog, popup

UserName

CAS RN 614-48-2

ONOO(E)

CAS RN 614-48-2

IUPAC Name: (2E)-3-(3-nitrophenyl)-1-phenylprop-2-en-1-one
Synonyms: chalcone, 3-nitro-
Molecular Weight (g/mol): 253.26
Molecular Formula: C15H11NO3
InChi Key: SMFBODMWKWBFOK-MDZDMXLPSA-N
SMILES: [O-][N+](=O)C1=CC=CC(\C=C\C(=O)C2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results